missing translation for 'onlineSavingsMsg'
Learn More

5-O-Benzyl-D-ribose, TRC

Product Code. 30179171
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179171 1 g 1g
30179049 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179171 Supplier Toronto Research Chemicals Supplier No. TRCB2888001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72369-89-2
Molecular Formula C12 H16 O5
Molecular Weight (g/mol) 240.25
IUPAC Name (2R,3R,4R)-2,3,4-trihydroxy-5-phenylmethoxypentanal
SMILES O[C@H](COCc1ccccc1)[C@@H](O)[C@@H](O)C=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H16O5/c13-6-10(14)12(16)11(15)8-17-7-9-4-2-1-3-5-9/h1-6,10-12,14-16H,7-8H2/t10-,11+,12-/m0/s1
Formula Weight 240.0998
Chemical Name or Material 5-O-Benzyl-D-ribose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.