missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)thymidine, 98+%

Product Code. 11440111
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
Unit Size:
0.1g
0.5g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440111 0.1 g 0.1g
11410111 0.5 g 0.5g
11420111 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440111 Supplier Thermo Scientific Alfa Aesar Supplier No. 044815.77
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40615-39-2 | C31H32N2O7 | 544.60 g/mol

5'-O-(4,4'-Dimethoxytrityl)thymidine is used in the solid-phase synthesis of polynucleotides and polythymidylic acids by the block coupling phosphotriester method. Further, it is used in the stereoselective synthesis of 3'-deoxy-3'-threo-hydroxymethyl nucleoside. In addition to this, it is used as a research tool for antiviral and anticancer studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40615-39-2
Molecular Formula C31H32N2O7
Molecular Weight (g/mol) 544.60
MDL Number MFCD00010113
InChI Key UBTJZUKVKGZHAD-QZGLRKMJNA-N
Synonym 5'-o-4,4'-dimethoxytrityl thymidine, dmt-t, thymidine, 5'-o-bis 4-methoxyphenyl phenylmethyl, 5'-o-4,4'-dimethoxytrityl thymidine dmt-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl-5-methylpyrimidine-2,4-dione, 5'-o-dimethoxytrityl-deoxythymidine, dmtr-dthd, dimethoxytrityl-t, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenyl-methoxy methyl-4-hydroxy-tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione
PubChem CID 162419
IUPAC Name 1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C)C(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Quantity 0.1 g
Formula Weight 544.6
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 5'-O- (4,4'-Dimethoxytrityl)thymidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.