missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine, 98%

Product Code. 15470397
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470397 Supplier Thermo Scientific Alfa Aesar Supplier No. J64880.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103285-22-9 | C31H32N2O8 | 560.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 103285-22-9
Color White to Yellow
Molecular Formula C31H32N2O8
MDL Number MFCD00274123
Quantity 1 g
Synonym 5'-o-4,4'-dimethoxytrityl-2'-o-methyluridine, 1-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxy-3-methoxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 5'-o-dmt-2'-o-methyl-uridine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-o-methyl-uridine, dmt-2-ome-u, uridine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-o-methyl, 5'-o-dmt-2'-o-methyluridine, mfdhavfjdsrpkc-yxinzvnlsa-n, 2'-o-methyl-5'-o-dmt-uridine, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-o-methyluridine
Solubility Information Soluble in acetonitrile
InChI Key MFDHAVFJDSRPKC-UHFFFAOYNA-N
Molecular Weight (g/mol) 560.60
PubChem CID 11039002
Formula Weight 560.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyluridine
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.