missing translation for 'onlineSavingsMsg'
Learn More

5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine, 98%

Product Code. 15457149
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15457149

Brand: Thermo Scientific Alfa Aesar J65960.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23669-79-6 | C30H30N2O7 | 530.58 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in acetonitrile at 20mg/ml

Notes
Store in freezer (-20°)
TRUSTED_SUSTAINABILITY

Specifications

CAS 23669-79-6
Color White
Molecular Formula C30H30N2O7
MDL Number MFCD00063410
Quantity 1 g
Synonym 5'-o-4,4'-dimethoxytrityl-2'-deoxyuridine, dmt-du, 2'-deoxy-5'-o-dmt-uridine, 2'-deoxy-5'-o-4,4'-dimethoxytrityl uridine, 1-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxyoxolan-2-yl-3h-pyrimidine-2,4-dione, 5'-o-bis 4-methoxyphenyl phenyl methyl-2'-deoxyuridine, uridine,5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, uridine, 5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy, 1-2r,4s,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-hydroxytetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 1-2r,4s,5r-5-bis 4-methoxyphenyl-phenylmethoxy methyl-4-hydroxyoxolan-2-yl pyrimidine-2,4-dione
Solubility Information Soluble in acetonitrile at 20mg/ml
InChI Key BYGKUWPLEGHFKX-ZRRKCSAHSA-N
SMILES COC1=CC=C(C=C1)C(OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
IUPAC Name 1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 530.58
PubChem CID 90230
Formula Weight 530.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.