missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo

Product Code. 10685002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10685002 100 g Glass bottle 100g
10341514 5 g Glass bottle 5g
10523121 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10685002 Leverantör Thermo Scientific Acros Leverantörsnummer 149101000
 Restricted Item
In Stock
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 5453-80-5 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 5453-80-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00167568
InChI Key AJIBZRIAUXVGQJ-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
PubChem CID 95117
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
SMILES C1C2CC(C1C=C2)C=O

Specifikationer

CAS Min % 90.0
CAS Max % 100.0
Density 1.0180g/mL
Boiling Point 67°C to 70°C (12.0 mmHg)
Flash Point 50.6°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.492
Quantity 100 g
Specific Gravity 1.018
Solubility Information Solubility in water: insoluble
Formula Weight 122.17
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Norbornene-2-carboxaldehyde, 90-95%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 5-Norbornene-2-carboxaldehyde, mixture of endo and exo
Signalord
  • Warning
Riskkategorier
  • Flammable liquid Category 3
Faroangivelser
  • H226-Flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.