missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxaldehyde, 95%, mixture of endo and exo

Product Code. 10341514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10341514 5 g Glass bottle 5g
10523121 25 g Glass bottle 25g
10685002 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10341514 Supplier Thermo Scientific Acros Supplier No. 149100050
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5453-80-5 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5453-80-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00167568
InChI Key AJIBZRIAUXVGQJ-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
PubChem CID 95117
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
SMILES C1C2CC(C1C=C2)C=O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Density 1.0180g/mL
Boiling Point 67°C to 70°C (12.0 mmHg)
Flash Point 50.6°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.492
Quantity 5 g
Specific Gravity 1.018
Solubility Information Solubility in water: insoluble
Formula Weight 122.17
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Norbornene-2-carboxaldehyde, 90-95%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Norbornene-2-carboxaldehyde, mixture of endo and exo
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.