missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornen-2-ol, mixture of endo and exo, 98+%

Product Code. p-7038100
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15463548

Brand: Thermo Scientific Alfa Aesar H56994.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13080-90-5 | C7H10O | 110.156 g/mol

It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

Solubility
Slightly soluble in water

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13080-90-5
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00167566
InChI Key MKOSBHNWXFSHSW-UHFFFAOYSA-N
Synonym 5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo
PubChem CID 96066
IUPAC Name bicyclo[2.2.1]hept-2-en-5-ol
SMILES C1C2CC(C1C=C2)O

Specifications

Melting Point 104°C to 106°C
Flash Point 51°C (124°F)
Quantity 1 g
UN Number UN1325
Solubility Information Slightly soluble in water
Formula Weight 110.15
Percent Purity 99%
Chemical Name or Material 5-Norbornen-2-ol, mixture of endo and exo

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.