missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanol, 97%, Thermo Scientific™

Product Code. 15225998 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15225998 25 mL 25mL
15215998 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15225998 Supplier Thermo Scientific Acros Supplier No. 390000250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 623-93-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
InChI Key FCBBRODPXVPZAH-UHFFFAOYSA-N
Synonym 5-nonanol, dibutylcarbinol, dibutyl carbinol, 5-nonyl alcohol, nonanol-5, acmc-1bc2t, di-n-butyl carbinol, 5-nonanol 25g, 5-nonanol gc, 5-nonanol, puriss gc
PubChem CID 12202
IUPAC Name nonan-5-ol
SMILES CCCCC(CCCC)O

Specifications

Melting Point 5.0°C
Density 0.8200g/mL
Boiling Point 194.0°C to 196.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3CH(OH)(CH2)2CH3
Refractive Index 1.429
Quantity 25 mL
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material 5-Nonanol
Show More Show Less
compliance-icons
Product Identifier
  • 5-Nonanol, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.