missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisatin, 97%

Product Code. 10031430 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031430 25 g Glass bottle 25g
10785401 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031430 Supplier Thermo Scientific Acros Supplier No. 128720250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-09-6 | C8H4N2O4 | 192.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-09-6
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005720
InChI Key UNMYHYODJHKLOC-UHFFFAOYSA-N
Synonym 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
PubChem CID 4669250
IUPAC Name 5-nitro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 251°C
Color Orange to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 456
Formula Weight 192.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroisatin, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Nitroisatin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.