missing translation for 'onlineSavingsMsg'
Learn More

5-nitrohexahydropyrimidine-2,4,6-trione hydrate, 97%, Thermo Scientific™

Product Code. 10055402
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10055402 10 g 10g
11366885 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055402 Supplier Thermo Scientific Maybridge Supplier No. RDP00108EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-58-7
Molecular Formula C4H5N3O6
Molecular Weight (g/mol) 191.10
MDL Number MFCD00160499
InChI Key YKGZAIJTCJOZNK-UHFFFAOYSA-N
Synonym 5-nitropyrimidine-2,4,6 1h,3h,5h-trione xhydrate, 5-nitrohexahydropyrimidine-2,4,6-trione hydrate, 5-nitro-1,3-diazinane-2,4,6-trione hydrate, 5-nitropyrimidine-2,4,6 1h,3h,5h-trione hydrate, 5-nitrobarbituric acid hydrate, c4nh33o5.h2o, 5-nitro-hexahydro-pyrimidine-2,4,6-trione, 2,4,6 1h,3h,5h-pyrimidinetrione,5-nitro-, hydrate 1:1
PubChem CID 2781274
SMILES O.[O-][N+](=O)C1C(=O)NC(=O)NC1=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name 5-nitro-1,3-diazinane-2,4,6-trione hydrate
Formula Weight 191.1
Percent Purity 97%
Chemical Name or Material 5-Nitrohexahydropyrimidine-2, 4, 6-trione hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.