missing translation for 'onlineSavingsMsg'
Learn More

(5-Nitrofuran-2-yl)methylene Diacetate, Mikromol™

Product Code. 30052691
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052691 25 mg 25mg
30052684 100 mg 100mg
30463233 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052691 Supplier Mikromol Supplier No. MM0943.020025
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(5-Nitrofuran-2-yl)methylene Diacetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (5-Nitrofuran-2-yl)methylene Diacetate
CAS 92-55-7
Analyte or Component Names (5-Nitrofuran-2-yl)methylene Diacetate
Shipping Condition +4°C
Molecular Formula C9 H9 N O7
Certifications/Compliance ISO 9001
Synonym Nifuroxazide Imp. C (EP), Nitrofural Imp. B (EP), Nitrofurfural Diacetate, (5-Nitrofuran-2-yl)methylidene Diacetate, Nifuroxazide Impurity C, Nitrofural Impurity B
InChI Formula InChI=1S/C9H9NO7/c1-5(11)15-9(16-6(2)12)7-3-4-8(17-7)10(13)14/h3-4,9H,1-2H3
SMILES CC(=O)OC(OC(=O)C)c1oc(cc1)[N+](=O)[O-]
IUPAC Name [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
Molecular Weight (g/mol) 243.17
Quantity 25 mg
Formula Weight 243.0379 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.