missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzimidazole, TRC

Product Code. 30470185
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30470185 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30470185

Brand: Toronto Research Chemicals TRCN4947951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Nitrobenzimidazole
CAS 94-52-0
Molecular Formula C7H5N3O2
Quantity 1 g
Synonym 1H-Benzimidazole','5-nitro- (9CI), Benzimidazole','5(or 6)-nitro- (6CI,7CI), Benzimidazole','5-nitro- (8CI), 5(6)-Nitrobenzimidazole, 5-Nitro-1H-benzimidazole, 5-Nitrobenzimidazole, 6-Nitro-1H-benzimidazole, 6-Nitrobenzimidazole, NSC 3068, NSC 58858
InChI Formula InChI=1S/C7H5N3O2/c11-10(12)5-1-2-6-7(3-5)9-4-8-6/h1-4H,(H,8,9)
SMILES [O-][N+](=O)c1ccc2nc[nH]c2c1
IUPAC Name 6-nitro-1H-benzimidazole
Molecular Weight (g/mol) 163.14
Formula Weight 163.0382
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.