missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-m-xylene, 99%

Product Code. 11322047
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322047 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11322047 Supplier Thermo Scientific Alfa Aesar Supplier No. L07762.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-12-7 | C8H9NO2 | 151.17 g/mol

5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-12-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007269
InChI Key BYFNZOKBMZKTSC-UHFFFAOYSA-N
Synonym 5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol
PubChem CID 7426
IUPAC Name 1,3-dimethyl-5-nitrobenzene
SMILES CC1=CC(=CC(C)=C1)[N+]([O-])=O

Specifications

Melting Point 69°C to 73°C
Boiling Point 273°C
Quantity 50 g
UN Number UN3447
Beilstein 1936130
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 5-Nitro-m-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.