missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-indanone, 97%, Thermo Scientific™

Product Code. 10215564
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215564 1 g Amber Glass Bottle 1g
10225564 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215564 Supplier Thermo Scientific Maybridge Supplier No. MO08378DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 116530-60-0
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00598938
InChI Key VSEBFWRYDORZJI-UHFFFAOYSA-N
PubChem CID 1502066
IUPAC Name 5-nitro-1,3-dihydroinden-2-one
SMILES [O-][N+](=O)C1=CC2=C(CC(=O)C2)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-nitro-2-indanone, 5-nitro-1h-inden-2 3h-one, 5-nitro-indan-2-one, 5-nitroindan-2-one, 2h-inden-2-one, 1,3-dihydro-5-nitro, 5-nitro-1,3-dihydro-2h-inden-2-one, pubchem18090, acmc-2099su, 2h-inden-2-one,1,3-dihydro-5-nitro, 1,3-dihydro-5-nitro-2h-inden-2-one
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.