missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furonitrile, 97%

Product Code. 11358455
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358455 25 g 25g
11348455 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358455 Supplier Thermo Scientific Alfa Aesar Supplier No. L10486.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 59-82-5 | C5H2N2O3 | 138.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-82-5
Molecular Formula C5H2N2O3
Molecular Weight (g/mol) 138.08
MDL Number MFCD00082647
InChI Key DUJNJLFQOODDNJ-UHFFFAOYSA-N
Synonym 5-nitro-2-furonitrile, 5-nitro-2-furancarbonitrile, unii-e63b56v8ya, 2-furancarbonitrile, 5-nitro, 5-nitro-2-cyanofuran, 5-nitro-2-furonitril, 2-nitrofuran-5-carbonitrile, acmc-1an56, 5-nitro-furan-2-carbonitrile, 2-furancarbonitrile,5-nitro
PubChem CID 94881
IUPAC Name 5-nitrofuran-2-carbonitrile
SMILES [O-][N+](=O)C1=CC=C(O1)C#N

Specifications

Melting Point 61°C to 64°C
Quantity 25 g
UN Number UN3439
Beilstein 123933
Formula Weight 138.08
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-furonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.