missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-2-furoic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Melting Point | 186°C to 188°C |
Quantity | 100 g |
Beilstein | 139373 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 157.08 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-Nitro-2-furoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'