missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furoic acid, 98+%

Product Code. 11433274
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433274 25 g 25g
11423274 5 g 5g
11443274 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433274 Supplier Thermo Scientific Alfa Aesar Supplier No. A10597.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 645-12-5 | C5H3NO5 | 157.081 g/mol

5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-12-5
Molecular Formula C5H3NO5
Molecular Weight (g/mol) 157.081
MDL Number MFCD00003240
InChI Key IODMEDPPCXSFLD-UHFFFAOYSA-N
Synonym 5-nitro-2-furoic acid, nitrofurate, 5-nitrofuroic acid, 2-furoic acid, 5-nitro, 5-nitro-2-furancarboxylic acid, 5-nitropyromucate, 5-nitrofurancarboxylic acid, 2-furancarboxylic acid, 5-nitro, ccris 1197, 5-nitropyromucic acid
PubChem CID 12577
IUPAC Name 5-nitrofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 186°C to 188°C
Quantity 25 g
Beilstein 139373
Solubility Information Slightly soluble in water.
Formula Weight 157.08
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-2-furoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.