missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldoxime, TRC

Product Code. 30475743
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 30475743

Brand: Toronto Research Chemicals TRCN493860250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nifuroxime
CAS 555-15-7
Molecular Formula C5 H4 N2 O4
Quantity 250 mg
Synonym 2-Furancarboxaldehyde','5-nitro-','oxime, 2-Furaldehyde','5-nitro-','oxime (6CI,7CI,8CI), 5-Nitro-2-furaldehyde oxime, 5-Nitro-2-furaldoxime, 5-Nitro-2-furancarboxaldehyde oxime, 5-Nitrofurfural oxime, 5-Nitrofurfuraldehyde oxime, Nifuroxime
InChI Formula InChI=1S/C5H4N2O4/c8-6-3-4-1-2-5(11-4)7(9)10/h1-3,8H
SMILES ON=Cc1oc(cc1)[N+](=O)[O-]
IUPAC Name N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
Molecular Weight (g/mol) 156.1
Formula Weight 156.0171
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.