missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldoxime, 97%, Thermo Scientific™

Product Code. 11354584
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354584 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11354584 Supplier Thermo Scientific Alfa Aesar Supplier No. L01374.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-15-7
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.10
MDL Number MFCD00198004,MFCD00198004
InChI Key PTBKFATYSVLSSD-ZZXKWVIFSA-N
Synonym nifuroxime, 5-nitro-2-furaldehyde oxime, usaf ea-5, 2-furaldehyde, 5-nitro-, oxime, 2-furaldoxime, 5-nitro, 5-nitro-2-furaldoxime, carbazylquinone, carbilquinone, 5-nitrofuran-2-carbaldehyde oxime, 5-nitro-2-furancarboxaldehyde oxime
PubChem CID 6478035
SMILES O\N=C\C1=CC=C(O1)[N+]([O-])=O

Specifications

Quantity 50 g
Beilstein 83037
Sensitivity Light sensitive
Merck Index 14,6536
IUPAC Name (NE)-N-[(5-nitrofuran-2-yl)methylidene]hydroxylamine
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material 5-Nitro-2-furaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.