missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-2-furaldehyde, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
Description
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
Specifications
Melting Point | 34°C to 38°C |
Density | 1.349 |
Boiling Point | 228°C |
Flash Point | 33°C (91°F) |
Refractive Index | 1.59 |
Quantity | 25 g |
Beilstein | 120539 |
Sensitivity | Air and light sensitive |
Solubility Information | Sparingly soluble in water. |
SMILES | C1=C(OC(=C1)[N+](=O)[O-])C=O |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'