missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde, 99%

Product Code. 11423324
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423324 50 g 50g
11394649 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423324 Supplier Thermo Scientific Alfa Aesar Supplier No. A10618.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 698-63-5 | C5H3NO4 | 141.082 g/mol

5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-63-5
Molecular Formula C5H3NO4
MDL Number MFCD00003230
InChI Key SXINBFXPADXIEY-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
PubChem CID 12774
ChEBI CHEBI:34459
IUPAC Name 5-nitrofuran-2-carbaldehyde

Specifications

Melting Point 34°C to 38°C
Density 1.349
Boiling Point 228°C
Flash Point 33°C (91°F)
Refractive Index 1.59
Quantity 50 g
Beilstein 120539
Sensitivity Air and light sensitive
Solubility Information Sparingly soluble in water.
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=O
Molecular Weight (g/mol) 141.082
Formula Weight 141.09
Percent Purity 99%
Chemical Name or Material 5-Nitro-2-furaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.