missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1H-pyrazole, 90%, Thermo Scientific™

Product Code. 10300953
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10300953 1 g Amber Glass Bottle 1g
10639744 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10300953 Supplier Thermo Scientific Maybridge Supplier No. FM00165DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 26621-44-3
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00159621,MFCD00238787
InChI Key MZRUFMBFIKGOAL-UHFFFAOYSA-N
Synonym 3-nitro-1h-pyrazole, 3-nitropyrazole, 1h-pyrazole, 3-nitro, 3-nitro-2h-pyrazole, nitropyrazole, pyrazole, 3-nitro, 5-nitro-1h-pyrazol, djw 94, djw-94, 5-nitropyrazole
PubChem CID 123419
IUPAC Name 5-nitro-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=NN1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 113.08
Percent Purity 97%
Chemical Name or Material 5-Nitro-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.