missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1H-indole-3-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10161082
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10161082 1 g 1g
10121182 10 g 10g
10756715 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10161082 Supplier Thermo Scientific Maybridge Supplier No. RH01174DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6625-96-3
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.158
MDL Number MFCD01313781
InChI Key PHKYMSLVWLYDKP-UHFFFAOYSA-N
Synonym 5-nitroindole-3-carbaldehyde, 5-nitroindole-3-carboxaldehyde, 5-nitro-1h-indole-3-carboxaldehyde, pubchem9126, 5-nitro-3-formylindol, 5-nitro-3-formyl-indole, 5-nitro-3-indolealdehyde, 5-nitroindole-3-aldehyde, 5-nitro-3-indolecarbaldehyde, 5-nitro-1h-indole-3-carb aldehyde
PubChem CID 246039
IUPAC Name 5-nitro-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=CN2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.16
Percent Purity 97%
Chemical Name or Material 5-Nitro-1H-indole-3-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.