missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1-benzothiophene-2-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10079351
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10079351 10 g Amber Glass Bottle 10g
10292832 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10079351 Supplier Thermo Scientific Maybridge Supplier No. MO00752EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 86010-32-4
Molecular Formula C9H4ClNO3S
Molecular Weight (g/mol) 241.65
MDL Number MFCD04115400
InChI Key KOJDFXDLYGKDEK-UHFFFAOYSA-N
Synonym 5-nitrobenzo b thiophene-2-carbonyl chloride, 5-nitrobenzothiophene-2-carboxoyl chloride, 5-nitro-1-benzo b thiophene-2-carbonyl chloride
PubChem CID 2794619
IUPAC Name 5-nitro-1-benzothiophene-2-carbonyl chloride
SMILES [O-][N+](=O)C1=CC=C2SC(=CC2=C1)C(Cl)=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 241.65
Percent Purity 95%
Chemical Name or Material 5-Nitro-1-benzothiophene-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.