missing translation for 'onlineSavingsMsg'
Learn More

5-(N,N-Hexamethylene)-amiloride, MedChemExpress

Product Code. 16478077
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478077 5 mg 5mg
16421557 50 mg 50mg
16405368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478077 Supplier Medchem Express Supplier No. HY128067
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-(N,N-Hexamethylene)-amiloride (Hexamethylene amiloride) derives from an amiloride and is a potent Na+/H+ exchanger inhibitor, which decreases the intracellular pH (pHi) and induces apoptosis in leukemic cells.

5-(N,N-Hexamethylene)-amiloride (Hexamethylene amiloride) is also an inhibitor of the HIV-1 Vpu virus ion channel and inhibits mouse hepatitis virus (MHV) replication and human coronavirus 229E (HCoV229E) replication in cultured L929 cells with EC50s of 3.91 μM and 1.34 μM, respectively.

Specifications

Chemical Name or Material 5-(N,N-Hexamethylene)-amiloride
CAS 1428-95-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C12H18ClN7O
Quantity 5 mg
Synonym Hexamethylene amiloride HMA
Solubility Information DMSO : 100 mg/mL (320.75 mM; Need ultrasonic)
SMILES O=C(C1=NC(Cl)=C(N2CCCCCC2)N=C1N)NC(N)=N
Molecular Weight (g/mol) 311.77
Formula Weight 311.77
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.