missing translation for 'onlineSavingsMsg'
Learn More

5-(N-Ethyl-N-isopropyl) Amiloride, TRC

Product Code. 30385353
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385353 5 mg
30388523 10 mg
30386317 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385353 Supplier Toronto Research Chemicals Supplier No. TRCE9215005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1154-25-2
Molecular Formula C11 H18 Cl N7 O
Molecular Weight (g/mol) 299.76
IUPAC Name 3-amino-N-carbamimidoyl-6-chloro-5-[ethyl(propan-2-yl)amino]pyrazine-2-carboxamide
SMILES CCN(C(C)C)c1nc(N)c(nc1Cl)C(=O)NC(=N)N

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H18ClN7O/c1-4-19(5(2)3)9-7(12)16-6(8(13)17-9)10(20)18-11(14)15/h5H,4H2,1-3H3,(H2,13,17)(H4,14,15,18,20)
Formula Weight 299.1261
Percent Purity >95%
Chemical Name or Material 5-(N-Ethyl-N-isopropyl) Amiloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.