missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10396514
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396514 1 g Amber Glass Bottle 1g
10789015 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396514 Supplier Thermo Scientific Maybridge Supplier No. CC73404DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 893744-01-9
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.28
MDL Number MFCD06803315
InChI Key YPXGCMYNDMFOHE-UHFFFAOYSA-N
Synonym 2-formyl-5-morpholin-4-ylmethyl thiophene, 5-morpholinomethyl thiophene-2-carbaldehyde, 5-morpholin-4-ylmethyl thiophene-2-carbaldehyde, 5-4-morpholinylmethyl-2-thiophenecarbaldehyde, 2-thiophenecarboxaldehyde,5-4-morpholinylmethyl, 5-morpholin-4-ylmethyl thiophene-2-carboxaldehyde, 5-morpholin-4-yl methyl thiophene-2-carbaldehyde
PubChem CID 20098933
IUPAC Name 5-(morpholin-4-ylmethyl)thiophene-2-carbaldehyde
SMILES O=CC1=CC=C(CN2CCOCC2)S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 211.28
Percent Purity 97%
Chemical Name or Material 5-(Morpholinomethyl)thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.