missing translation for 'onlineSavingsMsg'
Learn More

5-Morpholin-4-ylthiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10547484
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10547484 1 g Amber Glass Bottle 1g
10275804 250 mg Amber Glass Bottle 250mg
10422644 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10547484 Supplier Thermo Scientific Maybridge Supplier No. CC53404DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 24372-49-4
Molecular Formula C9H11NO2S
Molecular Weight (g/mol) 197.25
MDL Number MFCD02710611
InChI Key MWCGENRKAMIZLZ-UHFFFAOYSA-N
Synonym 5-morpholino-2-thiophenecarbaldehyde, 5-morpholinothiophene-2-carbaldehyde, 5-morpholin-4-yl thiophene-2-carbaldehyde, 5-morpholin-4-yl-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-4-morpholinyl, acmc-209gbl, morpholinothiophenecarbaldehyde, 5-morpholinothiophene-2-carboxaldehyde, 2-formyl-5-morpholin-4-yl thiophene, 5-morpholin-4-yl thiophene-2-carboxaldehyd
PubChem CID 2326306
IUPAC Name 5-morpholin-4-ylthiophene-2-carbaldehyde
SMILES O=CC1=CC=C(S1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.26
Percent Purity 97%
Chemical Name or Material 5-Morpholin-4-ylthiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.