missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KY 02111
GREENER_CHOICE

Product Code. 18706816
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18706816

Brand: Tocris Bioscience™ 4731/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits canonical Wnt signaling. Promotes differentiation of human ESCs and iPSCs into cardiomyocytes

KY 02111 promotes differentiation of human ESCs and iPSCs into cardiomyocytes when used after day three in culture; differentiation efficiency increases when used after initial culture with CHIR 99021 (Cat. No. 4423). Induces downregulation of Wnt signaling target genes; inhibits canonical Wnt signaling in a manner distinct from other known Wnt inhibitors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(6-Chloro-2-benzothiazolyl)-3,4-dimethoxybenzenepropanamide
CAS 1118807-13-8
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C18H17ClN2O3S
Formula Weight Observed MW: 376.86
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.