missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EMD 527040 hydrochloride

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
EMD 527040 hydrochloride is an inhibitor of integrin αVβ6. EMD 527040 inhibits binding of αVβ6 to fibronectin and attachment of cells expressing αVβ6 to fibronectin (IC50 values = 6 nM and 1.6 μM, respectively). EMD 527040 shows antifibrotic effects in an animal model of biliary fibrosis and is active in vivo.
Specifications
Specifications
| Chemical Name or Material | N-[1-Oxo-5-(2-pyridinylamino)pentyl]-O-(phenylmethyl)-L-seryl-3-(3,5-dichlorophenyl)-β-alanine hydrochloride |
| Quantity | 5 mg |
| Target | Integrin Receptor Inhibitors |
| Molecular Formula | C29H32Cl2N4O5.HCl |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction