missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CHS 828

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
CHS 828 is a NAMPT inhibitor. Decreases cellular level of NAD+. Selectively increases intracellular ROS levels in cancer cells but not normal cells. Active in vivo and cytotoxic.
Specifications
Specifications
| Chemical Name or Material | N-[6-(4-Chlorophenoxy)hexyl]-N'-cyano-N''-4-pyridinylguanidine |
| CAS | 200484-11-3 |
| Quantity | 5 mg |
| Molecular Formula | C19H22ClN5O |
| Formula Weight | 371.86 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction