missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ARC 239 dihydrochloride

Description
ARC 239 dihydrochloride is a selective α2B adrenoceptor antagonist (pKD values are 8.8, 6.7 and 6.4 at α2B, α2A, and α2D receptors respectively).
Specifications
Specifications
| Chemical Name or Material | 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione dihydrochloride |
| CAS | 55974-42-0 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C24H29N3O3.2HCl |
| Formula Weight | Observed MW: 480.43 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?