missing translation for 'onlineSavingsMsg'
Learn More

5-Methyluridine, 99%

Product Code. 15461369
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15461369 25 g 25g
15471369 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461369 Supplier Thermo Scientific Alfa Aesar Supplier No. H64259.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1463-10-1 | C10H14N2O6 | 258.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1463-10-1
Molecular Formula C10H14N2O6
Molecular Weight (g/mol) 258.23
MDL Number MFCD00006535
InChI Key DWRXFEITVBNRMK-JXOAFFINSA-N
Synonym 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine
PubChem CID 445408
ChEBI CHEBI:45996
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 183°C to 184°C
Color White
Quantity 25 g
Beilstein 90164
Formula Weight 258.23
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 5-Methyluridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.