missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5-Methyluridine

Product Code. 16403429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16403429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16403429 Supplier MedChemExpress Supplier No. HYW009444
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-Methyluridine is a is an endogenous methylated nucleoside found in human fluids.

Specifications

Molecular Weight (g/mol) 258.23 g/mol
Product Type Natural Products
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 97.0%
Quantity 1 g
Solubility Information H2O : 100 mg/mL (387.25 mM∣Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (387.25 mM)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 1463-10-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.