missing translation for 'onlineSavingsMsg'
Learn More

5-Methyltryptamine, hydrochloride, 98%, Thermo Scientific™

Product Code. 10542751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10542751 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10542751 Supplier Thermo Scientific Acros Supplier No. 158670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H15N2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00012683
InChI Key PYOUAIQXJALPKW-UHFFFAOYSA-O
Synonym 5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
PubChem CID 6917145
SMILES CC1=CC=C2NC=C(CC[NH3+])C2=C1

Specifications

CAS 1010-95-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 281°C to 283°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 2-(5-methyl-1H-indol-3-yl)ethanamine;hydrochloride
Formula Weight 210.71
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyltryptamine,hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.