missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carbonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 15487687
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487687 250 mg 250mg
15477687 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487687 Supplier Thermo Scientific Alfa Aesar Supplier No. H36784.MD
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31555-59-6
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.615
InChI Key DJCGIWVUKCNCAT-UHFFFAOYSA-N
Synonym 5-methyl-thiophene-2-carbonyl chloride, 5-methyl-2-thiophenecarbonyl chloride, 2-thiophenecarbonyl chloride, 5-methyl, 2-thiophenecarbonylchloride, 5-methyl, 5-methyl-2-thenoyl chloride, 2-chlorocarbonyl-5-methylthiophene, 5-methyl-thiophene-carboxylicacid chloride, 5-methylthiophene-2-carboxylic acid chloride, 5-methyl-thiophene-2-carboxylic acid chloride
PubChem CID 2760083
IUPAC Name 5-methylthiophene-2-carbonyl chloride
SMILES CC1=CC=C(S1)C(=O)Cl

Specifications

Color Colorless to Yellow
Boiling Point 78°C to 80°C (0.75 mmHg)
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture Sensitive
Formula Weight 160.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Methylthiophene-2-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.