missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carbonitrile, 97%, Thermo Scientific™

Product Code. 15254268 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254268 1 g 1g
15264268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254268 Supplier Thermo Scientific Acros Supplier No. 451010010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 72835-25-7
Molecular Formula C6H5NS
Molecular Weight (g/mol) 123.18
InChI Key RBQRZWYCXAXPIN-UHFFFAOYSA-N
PubChem CID 312733
IUPAC Name 5-methylthiophene-2-carbonitrile
SMILES CC1=CC=C(S1)C#N

Specifications

Boiling Point 73.0°C to 75.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 5-methyl-2-thiophenecarbonitrile, 2-cyano-5-methylthiophene, 2-thiophenecarbonitrile, 5-methyl, 2-thiophenecarbonitrile,5-methyl, 5-methyl-thiophene-2-carbonitrile
Formula Weight 123.18
Percent Purity 97%
Chemical Name or Material 5-Methylthiophene-2-carbonitrile
compliance-icons compliance-icons
Product Identifier
  • 5-Methylthiophene-2-carbonitrile, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.