missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carbonitrile, 95%, Thermo Scientific™

Product Code. 10294913
Change view
Click to view available options
Amber Glass Bottle; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10294913 Amber Glass Bottle; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10294913 Supplier Thermo Scientific Maybridge Supplier No. CC65016DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 72835-25-7
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C6H5NS
MDL Number MFCD01434334
Quantity 5g
Synonym 5-methyl-2-thiophenecarbonitrile, 2-cyano-5-methylthiophene, 2-thiophenecarbonitrile, 5-methyl, 2-thiophenecarbonitrile,5-methyl, 5-methyl-thiophene-2-carbonitrile
InChI Key RBQRZWYCXAXPIN-UHFFFAOYSA-N
SMILES CC1=CC=C(S1)C#N
IUPAC Name 5-methylthiophene-2-carbonitrile
Molecular Weight (g/mol) 123.173
PubChem CID 312733
Formula Weight 123.18
Percent Purity 95%
Chemical Name or Material 5-Methylthiophene-2-carbonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.