missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-boronic acid, 98%, ACROS Organics™

Product Code. 10038801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10038801 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10038801 Supplier Thermo Scientific Acros Supplier No. 359610010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 162607-20-7
CAS Min % 97.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H7BO2S
MDL Number MFCD01318166
Quantity 1 g
Synonym 5-methylthiophene-2-boronic acid, 5-methylthiophen-2-yl boronic acid, 5-methyl-2-thiopheneboronic acid, 5-methyl-2-thienyl boronic acid, 5-methyl-2-thienylboric acid, 5-methylthiophen-2-yl boranediol, 5-methyl-2-thienylboronic acid, 5-methylthiophene-2-boronicacid, 2-methylthiophene-5-boronic acid, 2-methylthienyl-5-boronic acid
InChI Key NRIYPIBRPGAWDD-UHFFFAOYSA-N
SMILES CC1=CC=C(S1)B(O)O
IUPAC Name (5-methylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 141.98
PubChem CID 2734374
Formula Weight 141.99
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 5-Methylthiophene-2-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.