missing translation for 'onlineSavingsMsg'
Learn More

5-Methyltetrahydrofolic acid, MedChemExpress

Product Code. 16435566
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16435566

Brand: Medchem Express HY113046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid. 5-Methyltetrahydrofolic acid is a methylated derivate of tetrahydrofolate. 5-Methyltetrahydrofolic acid is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid.

5-Methyltetrahydrofolic acid (5-Methyl THF) is a biologically active form of folic acid. 5-Methyltetrahydrofolic acid is a methylated derivate of tetrahydrofolate. 5-Methyltetrahydrofolic acid is the predominant natural dietary folate and the principal form of folate in plasma and cerebrospinal fluid.

Specifications

Chemical Name or Material 5-Methyltetrahydrofolic acid
Physical Form Solid
CAS 134-35-0
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H25N7O6
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym 5-Methyl THF
Solubility Information DMSO : 83.33 mg/mL (181.37 mM; Need ultrasonic)
SMILES O=C(O)CC[C@@H](C(O)=O)NC(C1=CC=C(NCC2N(C)C3=C(NC(N)=NC3=O)NC2)C=C1)=O
Molecular Weight (g/mol) 459.46
Formula Weight 459.46
Percent Purity 99.29%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.