missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylsulfonyl)thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-2702383
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10111282

Brand: Thermo Scientific Maybridge SEW03813CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 60166-86-1
Molecular Formula C6H6O4S2
Molecular Weight (g/mol) 206.23
MDL Number MFCD00173745
InChI Key SLWRTVINHWIGTK-UHFFFAOYSA-N
Synonym 5-methylsulfonyl thiophene-2-carboxylic acid, 5-methanesulfonylthiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 5-methylsulfonyl, maybridge1_007675, 5-methanesulfonyl-2-thiphenecarboxylic acid, 5-methylsulfonyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylicacid,5-methylsulfonyl, 5-methanesulfonyl thiophene-2-carboxylic acid
PubChem CID 2740497
IUPAC Name 5-methylsulfonylthiophene-2-carboxylic acid
SMILES CS(=O)(=O)C1=CC=C(S1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 5-(Methylsulfonyl)thiophene-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.