missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylpyrimidine, 98%, Thermo Scientific™
£58.90 - £166.00
Chemical Identifiers
CAS | 2036-41-1 |
---|---|
Molecular Formula | C5H6N2 |
Molecular Weight (g/mol) | 94.12 |
MDL Number | MFCD00047386 |
InChI Key | TWGNOYAGHYUFFR-UHFFFAOYSA-N |
Synonym | pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+% |
PubChem CID | 74859 |
IUPAC Name | 5-methylpyrimidine |
SMILES | CC1=CN=CN=C1 |
Description
Chemical Identifiers
2036-41-1 | |
94.12 | |
TWGNOYAGHYUFFR-UHFFFAOYSA-N | |
74859 | |
CC1=CN=CN=C1 |
C5H6N2 | |
MFCD00047386 | |
pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+% | |
5-methylpyrimidine |
Specifications
2036-41-1 | |
152°C to 154°C | |
97.5% min. (GC) | |
MFCD00047386 | |
Solubility in water: soluble | |
CC1=CN=CN=C1 | |
94.12 | |
94.11 | |
5-Methylpyrimidine |
33°C to 37°C | |
52°C | |
C5H6N2 | |
pyrimidine, 5-methyl, 5-methyl-pyrimidine, pyrimidine, 5-methyl-6ci,7ci,8ci,9ci, 5-methyl-pyrimidin, pubchem7207, rarechem am la 0031, 5-methyl-1,3-diazine, acmc-1cnd2, 5-methylpyrimidine, 5-methylpyrimidine, 97+% | |
TWGNOYAGHYUFFR-UHFFFAOYSA-N | |
5-methylpyrimidine | |
74859 | |
98% |
Safety and Handling
GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 218-006-9
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Methylpyrimidine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only