missing translation for 'onlineSavingsMsg'
Learn More

5-Methylnicotinic acid, 97%, Thermo Scientific™

Product Code. 15246038 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15246038 5 g 5g
15236038 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15246038 Supplier Thermo Scientific Acros Supplier No. 453670050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3222-49-9
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key DJDHHXDFKSLEQY-UHFFFAOYSA-N
Synonym 5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
PubChem CID 256208
IUPAC Name 5-methylpyridine-3-carboxylic acid
SMILES CC1=CN=CC(=C1)C(=O)O

Specifications

Melting Point 211.0°C to 213.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material 5-Methylnicotinic acid
compliance-icons
Product Identifier
  • 5-Methylnicotinic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.