missing translation for 'onlineSavingsMsg'
Learn More

5-Methylnicotinic acid, 97%

Product Code. 11359135
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359135 5 g 5g
11369135 250 mg 250mg
11349135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359135 Supplier Thermo Scientific Alfa Aesar Supplier No. L16370.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3222-49-9 | C7H7NO2 | 137.138 g/mol

5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylnicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Sparingly soluble (0.083 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3222-49-9
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00829036
InChI Key DJDHHXDFKSLEQY-UHFFFAOYSA-N
Synonym 5-methylnicotinic acid, 5-methyl-nicotinic acid, 5-methyl nicotinic acid, 5-methylnicotic acid, 5-methyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-methyl, 5-methyinicotinic acid, pubchem11961, 5-methyl nicotinic aicd, acmc-1afvp
PubChem CID 256208
IUPAC Name 5-methylpyridine-3-carboxylic acid
SMILES CC1=CN=CC(=C1)C(=O)O

Specifications

Melting Point 210°C to 215°C
Quantity 5 g
Beilstein 112288
Solubility Information Sparingly soluble (0.083g/L at 25°C).
Formula Weight 137.14
Percent Purity 97%
Chemical Name or Material 5-Methylnicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.