missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-3-carboxylic acid, 98+%

Product Code. 11388516
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11388516 Supplier Thermo Scientific Alfa Aesar Supplier No. L17577.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3405-77-4 | C5H5NO3 | 127.099 g/mol

It is a reactant for Click/aza-Michael or Click/oligomeric alkyl carbodiimide esterification reactions, preparation of aminopyrazole amide derivatives as Raf kinase inhibitors in melanoma cells and preparation of trifluoromethoxyphenyl(thiazolyl)pyrroles and other heterocycle-bearing polyfunctionalized pyrroles via chain heterocyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for Click/aza-Michael or Click/oligomeric alkyl carbodiimide esterification reactions, preparation of aminopyrazole amide derivatives as Raf kinase inhibitors in melanoma cells and preparation of trifluoromethoxyphenyl(thiazolyl)pyrroles and other heterocycle-bearing polyfunctionalized pyrroles via chain heterocyclization.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3405-77-4
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01318162
InChI Key BNMPIJWVMVNSRD-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
PubChem CID 76947
IUPAC Name 5-methyl-1,2-oxazole-3-carboxylic acid
SMILES CC1=CC(=NO1)C(=O)O

Specifications

Melting Point 171°C to 173°C
Quantity 5 g
Beilstein 114094
Formula Weight 127.1
Percent Purity ≥98%
Chemical Name or Material 5-Methylisoxazole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.