missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazol-3-amine, Mikromol™

Product Code. 30051858
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051858 25 mg 25mg
30049332 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051858 Supplier Mikromol Supplier No. MM0227.020025
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methylisoxazol-3-amine
TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material 5-Methylisoxazol-3-amine
CAS 1072-67-9
Analyte or Component Names 5-Methylisoxazol-3-amine
Shipping Condition Room Temperature
Molecular Formula C4 H6 N2 O
Certifications/Compliance ISO 17025
Synonym Sulfamethoxazole Imp. C (EP), Sulfamethoxazole USP Related Compound C, Sulfamethoxazole USP RC C, 5-Methylisoxazol-3-amine, Sulfamethoxazole Related Compound C, Sulfamethoxazole Impurity C
InChI Formula InChI=1S/C4H6N2O/c1-3-2-4(5)6-7-3/h2H,1H3,(H2,5,6)
SMILES Cc1onc(N)c1
IUPAC Name 5-methyl-1,2-oxazol-3-amine
Molecular Weight (g/mol) 98.1
Quantity 25 mg
Formula Weight 98.048 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.