missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindole, 99%

Product Code. 11377036
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11377036

Brand: Thermo Scientific Alfa Aesar L06893.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-96-0 | C9H9N | 131.178 g/mol

5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylindole is used as reactant for preparation of pharmaceutically active 2-oxo-1-pyrrolidine analogues, potential anticancer immunomodulators, IL2-inducible T-cell kinase (ITK) inhibitors and CRTh2 antagonists.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-96-0
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005680
InChI Key YPKBCLZFIYBSHK-UHFFFAOYSA-N
Synonym 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238
PubChem CID 11978
IUPAC Name 5-methyl-1H-indole
SMILES CC1=CC2=C(C=C1)NC=C2

Specifications

Melting Point 58°C to 61°C
Boiling Point 100°C (3 mmHg)
Quantity 1 g
Beilstein 2228
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 5-Methylindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.