missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindole, 99%, ACROS Organics™

Product Code. 10432401 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10432401

Brand: Thermo Scientific Acros 206850010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 614-96-0
CAS Min % 98.5
CAS Max % 100.0
Melting Point 58°C to 61°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H9N
MDL Number MFCD00005680
Quantity 1 g
Beilstein 20, 317
Synonym 5-methylindole, 1h-indole, 5-methyl, 5-methylindol, indole, 5-methyl, 5-methyl indole, unii-kt52363ri5, 5-methyl-indole, indole, 2, pubchem7238
InChI Key YPKBCLZFIYBSHK-UHFFFAOYSA-N
SMILES CC1=CC2=C(C=C1)NC=C2
IUPAC Name 5-methyl-1H-indole
Molecular Weight (g/mol) 131.178
PubChem CID 11978
Formula Weight 131.18
Percent Purity ≥98.5%
Physical Form Crystalline Solid
Chemical Name or Material 5-Methylindole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methylindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.