missing translation for 'onlineSavingsMsg'
Learn More

5-Methylindole-2-carboxylic acid, 99%

Product Code. 11383966
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383966 1 g 1g
11393966 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383966 Supplier Thermo Scientific Alfa Aesar Supplier No. L17145.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10241-97-1 | C10H9NO2 | 175.19 g/mol

5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylindole-2-carboxylic acid is a compound used in the synthesis of Pin1 inhibitors as potential antitumor agents. reactant in preparation of Pin1 inhibitors as potential antitumor agents reactant in preparation of non-imidazole human histamine H4 receptor antagonists

Solubility
Soluble in ethanol (50 mg/ml). Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10241-97-1
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00047166
InChI Key DAITVOCMWPNFTL-UHFFFAOYSA-N
Synonym 5-methylindole-2-carboxylic acid, 5-methyl-2-indolecarboxylic acid, chembl23328, 1h-indole-2-carboxylic acid, 5-methyl, acmc-1bxae, 5-me-ica, daitvocmwpnftl-uhfffaoysa, 5-methyl indole-2-carboxylic acid, 1h-indole-2-carboxylicacid, 5-methyl, #
PubChem CID 259188
IUPAC Name 5-methyl-1H-indole-2-carboxylic acid
SMILES CC1=CC=C2NC(=CC2=C1)C(O)=O

Specifications

Melting Point ∽235°C (decomposition)
Quantity 1 g
Beilstein 144055
Solubility Information Soluble in ethanol (50mg/ml). Insoluble in water.
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material 5-Methylindole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.