missing translation for 'onlineSavingsMsg'
Learn More

5-Methylimidazole-4-carboxaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11489027
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489027 5 g 5g
11479027 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489027 Supplier Thermo Scientific Alfa Aesar Supplier No. B20608.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68282-53-1 | Molecular Formula: C5H6N2O | Molecular Weight: 110.12 g/mol

4-Methyl-5-imidazolecarboxaldehyde is suitable for use in the synthesis of 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]-propan-2-ol (BIPO), 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]propane (BIP), {acetatoaqua[[(4-methylimidazol-5-yl)methylene]histamine]-copper(II)} perchlorate, Schiff base condensates with tris(2-aminoethyl)amine (tren) or tris(3-aminopropyl)amine (trpn). It is also used in the synthesis of imidazolate-based metal organic framework (MOF) membrane, 3,11-bis(4-imidazolylmethyl)-7-methyl-3,7,11,17-tetraazabicyclo[11.3.1]hepta-deca-1(17),13,15-triene, a 4-imidazolyl derivative of a 14-membered tetraazamacrocycle and Cu(II) eight and six coordinate compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-5-imidazolecarboxaldehyde is suitable for use in the synthesis of 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]-propan-2-ol (BIPO), 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]propane (BIP), {acetatoaqua[[(4-methylimidazol-5-yl)methylene]histamine]-copper(II)} perchlorate, Schiff base condensates with tris(2-aminoethyl)amine (tren) or tris(3-aminopropyl)amine (trpn). It is also used in the synthesis of imidazolate-based metal organic framework (MOF) membrane, 3,11-bis(4-imidazolylmethyl)-7-methyl-3,7,11,17-tetraazabicyclo[11.3.1]hepta-deca-1(17),13,15-triene, a 4-imidazolyl derivative of a 14-membered tetraazamacrocycle and Cu(II) eight and six coordinate compounds.

Solubility
Soluble in water, (23 g/L) (25°C).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68282-53-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00173728
InChI Key KMWCSNCNHSEXIF-UHFFFAOYSA-N
Synonym 4-methyl-1h-imidazole-5-carbaldehyde, 5-methylimidazole-4-carboxaldehyde, 4-methylimidazole-5-carboxaldehyde, 4-methyl-5-imidazolecarboxaldehyde, 5-methyl-3h-imidazole-4-carbaldehyde, 4-methylimidazole-5-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 5-methyl, 1h-imidazole-5-carboxaldehyde, 4-methyl, pubchem23016, acmc-1b6zs
PubChem CID 2795887
IUPAC Name 5-methyl-1H-imidazole-4-carbaldehyde
SMILES CC1=C(C=O)N=CN1

Specifications

Melting Point 166°C to 170°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in water,(23g/L) (25°C).
Formula Weight 110.12
Percent Purity 99%
Chemical Name or Material 5-Methylimidazole-4-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.